ContactUsLinkComponent
Formaway Formalin Neutralizing Absorption Pads, Mopec
  76579-376
 :  
undefined
Formaway Formalin Neutralizing Absorption Pads, Mopec
  76579-376
 :  BE092
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Formaway formalin neutralizing absorption pads
  • Size:
    3×4"
  • Environmentally Preferable:
  • Cat. No.:
    76579-376
Mopec’s Formaway Formalin Neutralizing Absorbent Pad (F-NAP) was designed to reduce formaldehyde vapors while dissecting tissue, transferring tissue from containers of formalin, and transporting or storing tissue in containers of formalin, by absorbing and neutralizing formalin.
  • F-NAP provides excellent vapor control during gross examination, dissecting, transporting, or transferring formalin fixed tissue
  • F-NAP Pads are also used for vapor control in storage of formalin, formalin fixed tissue containers and for small formalin spills
  • Blue color allows for easier viewing of small specimens, and the soft cutting surface allows for longer lasting scalpels
  • Sold in packs of 100
Mopec’s Formaway formalin neutralizing absorbent pad (F-NAP) was designed to facilitate the reduction of formaldehyde vapors by absorbing and neutralizing formaldehyde, glutaraldehyde and other aldehydes. Once the formalin solution is absorbed, the formalin neutralizing absorbent pad will convert formaldehyde and other aldehydes to a non-hazardous polymer, eliminating the source of harmful vapors making the work area safer.