Evergreen Slide Storage Boxes, Caplugs Evergreen
Supplier: Caplugs
Certificates
About this item
Caplugs Evergreen heavy-duty economical slide storage boxes feature a living hinge with one or two heavy-duty latches.
- Slide identification is made simple with a printed protective ID foam on the floor
- Boxes are stackable
Slides boxes are made from polypropylene.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.