Evergreen Forceps, Caplugs Evergreen
About this item
Caplugs Evergreen forceps are self-sprung and sturdy enough to be reusable, but economical enough to be disposable.
- Acetal (green): Resistant to solvents, acid and alkali
- ABS (blue): Features pointed end. Available in bulk packs and in individual sterile packs
- Polystyrene (transparent blue): Available in bulk packs and in individual sterile packs
5 Options Available Below
Product Details & Documents
Caplugs Evergreen forceps are self-sprung and sturdy enough to be reusable, but economical enough to be disposable.
- Acetal (green): Resistant to solvents, acid and alkali
- ABS (blue): Features pointed end. Available in bulk packs and in individual sterile packs
- Polystyrene (transparent blue): Available in bulk packs and in individual sterile packs
Product Family Options
Product Information
- MaterialColorLengthSterileAvailabilityPrice
- Supplier Number: 222-1121-B1I
- Supplier Number: 360-1120-B10
- Supplier Number: 360-1125-G90
- Supplier Number: 222-1130-B1I
- Supplier Number: 360-1129-B10