Dimethyl sulfoxide-[D₆] (99.95% D) for NMR spectroscopy
Supplier: VWR International
Certificates
About this item
CAS Number:2206-27-1
:
Formula:D₃CS(O)CD₃
MDL Number:MFCD00002090
:84.09 g/mol
Boiling Pt:≥173 … ≤209 °C (1,013 mbar)
Melting Pt:≥18 … ≤25 °C (1,013 mbar)
Density:1.19 g/cm³ (20 °C)
Flash Pt:88 °C (1 atm) (closed cup)
Storage Temperature:Ambient
EINECS:218-617-0
Specification Test Results
- Appearance/color:Colorless
- Appearance/form:Liquid
- D content 1H-NMR:min. 99.95 %(mol)
- w(H2O) 1H-NMR:max. 0.010 %(w)
- 1H-NMR Purity:conform
- H2O + D2O Content by KF Coulometer:max 0.020 %(w)
- GC Content:min. 99.5 %(w)
Specification Test Results
Frequently Bought Together