

Dichloromethane-[D₂] (99.8% D) for NMR spectroscopy
Supplier: BDH
Certificates
About this item
CAS Number:1665-00-5
:
Formula:CD₂Cl₂
MDL Number:MFCD00000882
:86.92 g/mol
Boiling Pt:39 … 40 °C
Melting Pt:–95 °C (101 kPa)
Density:1.36 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-776-0
UN:1593
ADR:6.1,III
:GHS07
:GHS08
Specification Test Results
- Appearance/color:Colorless
- Appearance/form:Liquid
- D content 1H-NMR:min. 99.80 %(mol)
- w(H2O) 1H-NMR:max. 0.010 %(w)
- 1H-NMR Purity:conform
- H2O + D2O Content by KF Coulometer:max 0.020 %(w)
- GC Content:min. 99.0 %(w)
Specification Test Results
Frequently Bought Together