

Chloroform-[D₁] (99.8% D) + 0.03% TMS for NMR spectroscopy
Supplier: VWR International
Certificates
About this item
CAS Number:865-49-6
:
Formula:CDCl₃
MDL Number:MFCD00000827
:120.38 g/mol
Boiling Pt:61.5 °C (1013 hPa)
Melting Pt:–64.7 … –64.2 °C (4 hPa)
Density:1.45 g/cm³ (20 °C)
Flash Pt:> 60 °C (1019 hPa)
Storage Temperature:Refrigerator
EINECS:212-742-4
UN:1888
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance/color:Colorless
- Appearance/form:Liquid
- D content 1H-NMR:min. 99.80 %(mol)
- w(H2O) 1H-NMR:max. 0.010 %(w)
- 1H-NMR Purity:conform
- H2O + D2O Content by KF Coulometer:max 0.020 %(w)
- v(TMS) 1H-NMR:0.02-0.04 %V)
Specification Test Results
Frequently Bought Together