Evergreen Inoculum Trays, Caplugs Evergreen
: Caplugs
Caplugs Evergreen precision molded 95-pin inoculator can simultaneously pick up 95 bacterial colony samples (10 µl/pin) and transfer them into or from a 96-well format microplate or microplate tray. The missing pin in the inoculator assembly serves as a blank control.
- Ideal for antimicrobial susceptibility testing, urine cultures, bacterial colony subcultures, cell/tissue cultures, cytotoxicity testing, serology dilution studies and evaluation of growth media
- Applications include cell/tissue culture, blood toxicity and serological studies or other dilution and sensitivity tests
- Both the inoculator assembly and the inoculum tray are gamma sterilized
Each 95-pin assembly comes pre-assembled with a 40 ml inoculum tray. Additional trays are also available separately.
Frequently Bought Together