Evergreen Centrifuge Tubes, 15 ml, Caplugs Evergreen
Supplier: Caplugs
Certificates
About this item
Caplugs Evergreen 15 ml Centrifuge Tubes are precision molded from premium-grade, chemically-inert thermoplastics. Polystyrene tubes are glass-clear and polypropylene is nearly transparent.
- Polystyrene tubes withstand up to 6200×g and are appropriate for use with mild acidic (except oxidizing acids) or alkali solutions, alcohols (except higher alcohols) and salt
- Polypropylene tubes withstand up to 12100×g and are appropriate for use with stronger acids (except oxidizing acids) or alkalis, alcohols, organic solvents, chlorinated and aromatic hydrocarbons (except xylene) and ketones
- Screw caps have molded-in sealing rings and heavy-duty threads, the quality assurance program features vacuum chamber testing (95 Kpa) to ensure that these caps close tightly every time
Accurate, easy-to-read graduations are molded-in at 0.1 ml intervals from 0 to 1 ml and at 0.5 ml intervals from 1 to 5 ml and every 1 ml from 5 to 15 ml. The outside diameter is 17 mm and the overall length is 120 mm.
Packaging: Sterile tubes with screwcaps are packaged in reclosable 50-tube tray packs (10 trays/case) or in 25-tube bags (20 bags/case). Non-sterile tubes with screwcaps are sold in 1000-count bulk units.
Frequently Bought Together