Klercide™ Cleanroom Quat Metered Dose Concentrate Non-Sterile
Catalog # 76554-248
Supplier: Avantor
Specifications
- Description:Single-bagged
- Size:4x1 gal
- Cat. No.:76554-248
- Packaging:Bottle, capped
- Sterility:Non sterile
- Form:Metered dose concentrate
Specifications
About this item
Cleanroom Quat Metered Dose Concentrate Non-Sterile is a blend of quaternary ammonium compound and deionized water. Concentrated non-food contact sanitizer, disinfectant, and fungicide for hard, non-porous surfaces for use in cleanrooms and controlled areas.
- Is easy and economical to use
- Is 0.2 micron filtered, filled and single bagged in a cleanroom environment
- Non-corrosive for use in most applications
- Compatible with other Ecolab biocides for rotational use
Each 1-gallon container makes 50 gallons of diluted sanitizer (76 ml of concentrate makes 1 gallon of ready-to-use sanitizer). Flexible use instructions allow for product use as a sanitizer, disinfectant, or fungicidal application.
EPA Registered Sanitizer, Disinfectant, and Fungicide.
EPA Reg. # 1677-264