ContactUsLinkComponent
Evergreen Two Slide Mailers, Caplugs Evergreen
Evergreen Two Slide Mailers, Caplugs Evergreen
Catalog # 76520-586
Supplier:  Caplugs
undefined
Evergreen Two Slide Mailers, Caplugs Evergreen
Catalog # 76520-586
Supplier:  Caplugs
Supplier Number:  240-3072-030
Specifications
  • Color:
    Natural
  • Description:
    2-Compartment slide mailer, polypropylene
  • Cat. No.:
    76520-586
Specifications
About this item
Caplugs Evergreen two-slide mailers feature a sturdy living hinge design with friction-fit and post-type locks.
  • Top-panel window allows slides to be viewed without opening
  • Easy-to-use finger/thumb tabs eliminate prying and/or cutting tape
  • Available in multiple colors
A large matte-finished area is molded-in to provide ample room for identification. The slim shape makes it ideal as a component of transport and mailing kits.