Evergreen One Slide Mailers, Caplugs Evergreen
Supplier: Caplugs
Certificates
About this item
Caplugs Evergreen one-slide mailers feature a sturdy living hinge design with friction-fit and post-type locks.
- Top-panel window allows slides to be viewed without opening
- Easy-to-use finger/thumb tabs eliminate prying and/or cutting tape
- Available in multiple colors
A large matte-finished area is molded-in to provide ample room for identification. The slim shape makes it ideal as a component of transport and mailing kits.
Frequently Bought Together