ContactUsLinkComponent
VWR® Performance Laboratory Freezers with Manual Defrost (30 cu. ft.)
VWR® Performance Laboratory Freezers with Manual Defrost (30 cu. ft.)
Supplier:  VWR International
Certificates
undefined
VWR® Performance Laboratory Freezers with Manual Defrost (30 cu. ft.)
Supplier:  VWR International
Specifications
  • Catalog No:
  • 76580-198
  • 76580-200
  • Cat. No.:
  • 76580-198
  • 76580-200
  • Defrost:
  • Manual
  • Manual
  • Electrical:
  • 115 V, 60 Hz, 10.7 A, 1 HP
  • 115 V, 60 Hz, 6 A, ⅓ HP
  • Shelves/drawers:
  • 4 fixed shelves
  • 4 fixed shelves
  • Door Type:
  • 1 solid door, right hinged
  • 1 solid door, right hinged
  • Weight:
  • 210 kg (462 lbs.)
  • 210 kg (462 lbs.)