VWR® Standard Hazardous Location (Explosion Proof) Refrigerator Freezer Combo Unit with Manual Defrost (30 cu. ft.)
Catalog # 76580-130
Supplier: VWR International
Specifications
- Temperature range:1...10 °C Refrigerator / ‒15...‒25 °C Freezer
- Capacity:850 L (30 cu.ft.)
- Description:Standard series laboratory refrigerator/freezer combo hazardous location (explosion proof), 30 CF
- Cat. No.:76580-130
- Electrical:115 V, 60 Hz, 6 A, ⅓ HP
- Defrost:Cycle (Refrigerator) / Manual (Freezer)
- Door Type:1 solid door, right hinged
- Shelves/drawers:3 adjustable shelves (refrigerator)
- Weight:185 kg (408 lbs.)
- Exterior W×D×H:908×921×1969 mm (35¾×36¼×77½")
- Interior W×D×H:737×565×305 mm (29×22¼×12") Freezer / 813×572×1283 mm (32×22½×50½") Refrigerator
Specifications
About this item
They utilize non-sparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist. For storage of flammable or volatile materials within typical laboratory or non-volatile environments, see flammable material storage units.
- Mechanical thermostat sealed in explosion proof box
- Keyed door lock
- High density foam insulation
- Must be hardwired for safety
These units are equipped with interior shelving, and microprocessor temperature controllers. The standard line provides multi-functional features in a cost-effective design.
Material: White powder coat
Certifications: These units meet NFPA and OSHA guidelines for hazardous location, Class 1, Division II, Groups C and D guidelines.
Ordering information: Units come with a two year parts and labor warranty.
Caution: Units are not equipped with a line cord and plug. They must be hardwired in conduit to 120 VAC power source for safety.