ContactUsLinkComponent
Plastic Connectors 25 mm TC - Threaded, cytiva
Plastic Connectors 25 mm TC - Threaded, cytiva
Catalog # 104013-252
Supplier:  Cytiva
undefined
Plastic Connectors 25 mm TC - Threaded, cytiva
Catalog # 104013-252
Supplier:  Cytiva
Supplier Number:  18117008
Specifications
  • Description:
    Plastic connectors 25 mm TC - threaded
  • Cat. No.:
    104013-252
  • Thread size:
    ¹/₄"
Specifications
About this item
These connectors allow for connection from a 25 mm TriClamp (TC) connector to various threaded connectors.