Order Entry
United States
ContactUsLinkComponent
HiTrap™ Macrocap SP Columns, Cytiva
HiTrap™ Macrocap SP Columns, Cytiva
Catalog # 104013-114
Supplier:  Cytiva
HiTrap™ Macrocap SP Columns, Cytiva
Catalog # 104013-114
Supplier:  Cytiva
Supplier Number:  28950859

Specifications

  • Capacity:
    5 ml
  • Description:
    HiTrap™ MacroCap SP
  • Storage:
    4 ...30 °C, 20% Ethanol, 0.2 M Sodium Acetate
  • Bed dimensions:
    16×25 mm
  • Cat. No.:
    104013-114

Specifications

About this item