Order Entry
United States
ContactUsLinkComponent
HiLoad™ with Superdex™ Prep Grade Gel Filtration Chromatography Columns
HiLoad™ with Superdex™ Prep Grade Gel Filtration Chromatography Columns
Supplier:  Cytiva
Certificates
undefined
HiLoad™ with Superdex™ Prep Grade Gel Filtration Chromatography Columns
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 76488-170
  • 76488-172
  • 104013-194
  • Max. Pressure:
  • 3 bar
  • 3 bar
  • 3 bar
  • pH Range:
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4...30 °C, 20% ethanol
  • 4...30 °C, 20% ethanol
  • 4...30 °C, 20% ethanol

Specifications

Frequently Bought Together