Human Recombinant respiratory syncytial virus (RSV) (A, strain Long) Fusion glycoprotein F0/RSV-F Protein (His Tag) (from Baculovirus Insect Cells)
Catalog # 76535-118
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:RSV
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:P12568
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:fusion protein|HRSVgp08|F
- Protein/Peptide Name:RSV-F
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant Human respiratory syncytial virus (RSV) (A, strain Long) Fusion glycoprotein F0 consists of 510 amino acids and predicts a molecular mass of 56.92 kDa. As a result of glycosylation, it migrates as an approximately 75.98 and 61.80 KDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Asn524
- Shipping Temperature:Ambient
- Cat. No.:76535-118
Specifications
About this item
A DNA sequence encoding the human respiratory syncytial virus (RSV) (A, strain Long) fusion glycoprotein F0 (P12568) (Met1-Asn524) was expressed with a polyhistidine tag at the C-terminus.