Human Recombinant coronavirus HKU1 (isolate N5) (HCoV-HKU1) Spike Protein (S1+S2 ECD, His Tag) (from Baculovirus Insect Cells)
Catalog # 103824-996
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:HCoV-HKU1(isolate N5)
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:Q0ZME7.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ncov RBD|ncov s2|ncov spike|coronavirus s2|S|s1|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|RBD
- Protein/Peptide Name:Spike
- Purity:>75% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human coronavirus HKU1 (isolate N5) (HCoV-HKU1) spike protein (S1+S2 ECD, His Tag) consists of 1294 amino acids and predicts a molecular mass of 144.4 kDa.
- Sequence:Met1-Pro1295
- Shipping Temperature:Ambient
- Cat. No.:103824-996
Specifications
About this item
A DNA sequence encoding the human coronavirus HKU1 (isolate N5) (HCoV-HKU1) spike protein (S1+S2 ECD) (Q0ZME7.1) (Met1-Pro1295) was expressed with a polyhistidine tag at the C-terminus.