HOBO® 4-Channel Analog Data Logger
Catalog # 470323-530
Supplier: ONSET COMPUTER CORPORATION SE
Specifications
- Data Memory:4 MB (1.9 million measurements, max.)
- Description:HOBO® 4-channel analog data logger
- Battery Life:1 year, typical with logging rate of 1 min
- Alarm:Yes
- Cat. No.:470323-530
Specifications
About this item
This logger supports up to four external sensors to record parameters like temp, current, etc.
- Twice the accuracy over previous models with 16-bit resolution
- Flexible support for a wide range of external sensors
- LCD confirms logger operation and displays near real-time measurement data
- Provides minimum, maximum, average, and standard deviation logging options
- On-screen alarms notify you when a sensor reading exceeds set thresholds
- Stores 1.9 million measurements for longer deployments between offloads
- Compatible with HOBOware® and HOBOware® Pro software for logger setup, graphing, and analysis
A high-performance, LCD display data logger, supporting up to four external sensors to measure and record parameters such as temperature, current, CO₂, and voltage, for building performance monitoring applications. This logger is ideal for energy audits, building commissioning studies, equipment scheduling, and more.
Ordering information: Requires HOBOware® software and a USB cable. (Note: A USB cable is included with HOBOware® Pro CD or USB software purchase). See compatible items below.