Mouse Heparin-Binding Epidermal Growth Factor-like Growth Factor (HB-EGF) ELISA Kit, AFG Bioscience
76578-320
:
- Assay Type:Sandwich
- Host:Mouse
- Target protein:—
- Description:Mouse heparin-binding epidermal growth factor-like growth factor,HB-EGF ELISA kit
- Size:96 Wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:3.3 - 200 pg/ml
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 µl
- Sensitivity:0.8 pg/ml
- Regulatory Status:RUO
- Cat. No.:76578-320
- Assay Principle: Quantitative
- Research Topic: Cardiovascular
- Detection Wavelength: 450 nm