Order Entry
United States
ContactUsLinkComponent
Mouse C-Reactive Protein (CRP) ELISA Kit, AFG Bioscience
Mouse C-Reactive Protein (CRP) ELISA Kit, AFG Bioscience
Catalog # 76576-950
Supplier:  AFG Bioscience
CAS Number:  
undefined
Mouse C-Reactive Protein (CRP) ELISA Kit, AFG Bioscience
Catalog # 76576-950
Supplier:  AFG Bioscience
Supplier Number:  EK730110
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Host:
    Mouse
  • Target protein:
    C-Reactive Protein
  • Description:
    Mouse C-Reactive Protein ELISA kit
  • Size:
    96 Wells
  • Sample Type:
    Serum, plasma, urine, tissue homogenates, cell culture supernates
  • Time to Results:
    1 - 5 h
  • Shelf Life:
    6 months (4 °C); 12 months (–20 °C)
  • Detection Range:
    33 - 2000 pg/ml
  • Storage Temperature:
    2…8 °C
  • Sample Volume:
    50 - 100 µl
  • Sensitivity:
    8 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76576-950

Specifications

About this item

  • Assay Principle: Quantitative
  • Research Topic: Immunology
  • Detection Wavelength: 450 nm