Order Entry
United States
ContactUsLinkComponent
Mouse Complement Fragment 3a (C3a) ELISA Kit, AFG Bioscience
Mouse Complement Fragment 3a (C3a) ELISA Kit, AFG Bioscience
Catalog # 76578-316
Supplier:  AFG Bioscience
CAS Number:  
undefined
Mouse Complement Fragment 3a (C3a) ELISA Kit, AFG Bioscience
Catalog # 76578-316
Supplier:  AFG Bioscience
Supplier Number:  EK730104
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Host:
    Mouse
  • Target protein:
    Complement fragment 3a,C3a
  • Description:
    Mouse Complement fragment 3a,C3a ELISA kit
  • Size:
    96 Wells
  • Sample Type:
    Serum, plasma, urine, tissue homogenates, cell culture supernates
  • Time to Results:
    1 - 5 h
  • Shelf Life:
    6 months (4 °C); 12 months (–20 °C)
  • Detection Range:
    3 - 200 pg/ml
  • Storage Temperature:
    2…8 °C
  • Sample Volume:
    50 - 100 µl
  • Sensitivity:
    0.5 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76578-316

Specifications

About this item

  • Assay Principle: Quantitative
  • Research Topic: Cancer
  • Detection Wavelength: 450 nm