Mouse Adrenomedullin (ADM) ELISA Kit, AFG Bioscience
Catalog # 76576-474
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:Adrenomedullin,ADM
- Description:Mouse adrenomedullin,ADM ELISA kit
- Size:96 Wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:3 - 180 pg/ml
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 µl
- Sensitivity:0.5 pg/ml
- Regulatory Status:RUO
- Cat. No.:76576-474
Specifications
About this item
- Assay Principle: Quantitative
- Research Topic: Signal Transduction
- Detection Wavelength: 450 nm