Rat Interleukin-19 (IL-19) ELISA Kit, AFG Bioscience
Catalog # 76576-152
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Rat
- Target protein:Interleukin 19
- Description:Rat Interleukin 19(IL-19) ELISA kit
- Size:96 Wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:-
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 µl
- Sensitivity:-
- Regulatory Status:RUO
- Cat. No.:76576-152
Specifications
About this item
- Research Topic: Immunology
- Abbreviation: IL-19
- Detection Wavelength: 450 nm