Fixative Bouins, Statlab
Supplier: StatLab
Certificates
About this item
Saturated aqueous picric acid/formalin solution.
- Bouins is commonly used in trichrome staining procedures and with GI tract specimens
Packaging: Manufactured in the USA