Rat Insulin-like Growth Factor 2 mRNA-Binding Protein 3 (IGF2BP3) ELISA Kit, AFG Bioscience
Catalog # 76578-662
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Rat
- Target protein:Insulin-like growth factor 2 mRNA-binding protein 3
- Description:Rat insulin-like growth factor 2 mRNA-binding protein 3 (IGF2BP3) ELISA kit
- Size:96 Wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:-
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 µl
- Sensitivity:-
- Regulatory Status:RUO
- Cat. No.:76578-662
Specifications
About this item
- Research topic: Epigenetics and nuclear signaling, signal transduction
- Abbreviation: IGF2BP3
- Detection wavelength: 450 nm