Rat 1, 25- Hydroxy Vitamin D3 (1, 25 (OH) 2 D3) ELISA Kit, AFG Bioscience
Catalog # 76578-410
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Rat
- Target protein:1,25- hydroxy vitamin D3
- Description:Rat 1,25- hydroxy vitamin D3 (1,25(OH) 2 D3) ELISA kit
- Size:96 Wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:-
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 µl
- Sensitivity:-
- Regulatory Status:RUO
- Cat. No.:76578-410
Specifications
About this item
- Research topic: Metabolism
- Abbreviation: 1,25(OH) 2 D3
- Detection wavelength: 450 nm