Order Entry
United States
ContactUsLinkComponent
Human Tryptophan (Trp) ELISA Kit, AFG Bioscience
Human Tryptophan (Trp) ELISA Kit, AFG Bioscience
Catalog # 76577-976
Supplier:  AFG Bioscience
CAS Number:  
undefined
Human Tryptophan (Trp) ELISA Kit, AFG Bioscience
Catalog # 76577-976
Supplier:  AFG Bioscience
Supplier Number:  EK711758
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Host:
    Human
  • Target protein:
    Tryptophan
  • Description:
    Human Tryptophan (Trp) ELISA kit
  • Size:
    96 Wells
  • Sample Type:
    Serum, plasma, urine, tissue homogenates, cell culture supernates
  • Time to Results:
    1 - 5 h
  • Shelf Life:
    6 months (4 °C); 12 months (–20 °C)
  • Detection Range:
    0.2 - 10 nmol/ml
  • Storage Temperature:
    2…8 °C
  • Sample Volume:
    50 - 100 µl
  • Sensitivity:
    0.05 nmol/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76577-976

Specifications

About this item

  • Assay principle: Quantitative
  • Abbreviation: Trp
  • Detection wavelength: 450 nm