Human Platelet-Derived Growth Factor-BB (PDGF-BB) ELISA Kit, AFG Bioscience
Catalog # 76576-306
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:Platelet-Derived Growth Factor-BB
- Description:Human platelet-derived growth factor-BB (PDGF-BB) ELISA kit
- Size:96 Wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:4.6 - 300 pg/ml
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 µl
- Sensitivity:1.2 pg/ml
- Regulatory Status:RUO
- Cat. No.:76576-306
Specifications
About this item
- Assay principle: Quantitative
- Research topic: Cancer, cardiovascular
- Abbreviation: PDGF-BB
- Detection wavelength: 450 nm