ContactUsLinkComponent
Heating Cords (HTC) With SDC Temperature Controller Bundle, BriskHeat
Heating Cords (HTC) With SDC Temperature Controller Bundle, BriskHeat
Supplier:  BRISKHEAT MS
Certificates
Heating Cords (HTC) With SDC Temperature Controller Bundle, BriskHeat
Supplier:  BRISKHEAT MS
Specifications
  • Catalog No:
  • MSPP-UX3612019
  • MSPP-HTC451003
  • Min. operating temperature:
  • 32 °F (0 °C)
  • 32 °F (0 °C)
  • Max. Operating Temperature:
  • 900 °F (482 °C)
  • 900 °F (482 °C)
  • Warranty:
  • 1 year
  • 1 year