NEBuffer r2.1, New England Biolabs
Catalog # 76559-402
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Concentration:10X
- Description:NEBuffer r2.1
- Size:5 ml
- Cat. No.:76559-402
Specifications
About this item
New England Biolabs provides a color-coded 10 X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. These buffers contain Recombinant Albumin (NEB #B9200).
- Contains recombinant Albumin
- Contains four 1.25 ml vials of NEBuffer r2.1