Order Entry
United States
ContactUsLinkComponent
NEBuffer r3.1, New England Biolabs
NEBuffer r3.1, New England Biolabs
Catalog # 76559-396
CAS Number:  
undefined
NEBuffer r3.1, New England Biolabs
Catalog # 76559-396
Supplier Number:  B6003S
CAS Number:  

Specifications

  • Concentration:
    10X
  • Description:
    NEBuffer r3.1
  • Size:
    5 ml
  • Cat. No.:
    76559-396

Specifications

About this item

New England Biolabs provides a color-coded 10 X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. These buffers contain Recombinant Albumin (NEB #B9200).

  • Contains recombinant Albumin
  • Contains four 1.25 ml vials of NEBuffer r3.1