Order Entry
United States
ContactUsLinkComponent
Eisco Small Beaker Tongs
Eisco Small Beaker Tongs
Catalog # 470345-918
Supplier:  Wards
undefined
Eisco Small Beaker Tongs
Catalog # 470345-918
Supplier:  Wards
Supplier Number:  BKTGMINI

Specifications

  • Description:
    Small beaker tongs
  • Environmentally Preferable:
  • Cat. No.:
    470345-918
  • W×L:
    2.5×7.25"

Specifications

About this item

Ideal for gentle handling of smaller sized glassware.

  • Handle made of high-quality, nickel-plated steel
  • Curved jaws with rubber coated tips for added grip strength and safety
  • For use with 25 to 100 ml capacity beakers

Small metal beaker tongs made of nickel-plated steel. Tongs measure 7.25" in length and are 2.5" wide.