Micro Tip Boxes, United Scientific Supplies
Supplier: United Scientific Supplies
Certificates
About this item
Micro tip storage boxes.
- Holes are uniformly spaced for use with a multi-channel micropipette
- Transparent top covers included
- Autoclavable
Pack of six empty micro tip boxes; each box holds up to 96 tips with 2 to 200 μl capacity. Holes are uniformly spaced for use with a multi-channel micropipette.
Frequently Bought Together