Order Entry
United States
ContactUsLinkComponent
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane 97%
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane 97%
Catalog # TS43412-1000
CAS Number:  
undefined
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane 97%
Catalog # TS43412-1000
Supplier Number:  43412-1000
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 mg
  • Cat. No.:
    TS43412-1000

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • NMR:
    ≥96.0 %
  • GC:
    ≥99.5 %
  • Residue after evaporation:
    ≤0.001 %

Specification Test Results

About this item

 :
 :292.36 g/mol
Flash Pt:>110 °C
Storage Temperature:Ambient
CAS Number:1314246-02-0
Formula:C₁₇H₂₅O₂P
MDL Number:MFCD09265099