Eosin, Select
Supplier: StatLab
Jump to:
About this item
Proprietary, alcoholic and acidified formulation.
- Produces a brilliant cytoplasmic stain with three distinct color variations in a short amount of staining time
- Yields a very familiar cytoplasmic staining result
Part of the StatLab Select stain line.
Manufactured in USA.
Synonyms: Acid Red 87, Tetrabromofluorescein, 2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acid, 2,4,5,7-Tetrabromofluorescein, Eosin yellowish, Solvent red 43, Bromofluorescein, Bromo acid J. TS, XL, or XX, CI 45380, Bronze Bromo ES, Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-
- CAS Number:15086-94-9
- Formula:C₂₀H₈Br₄O₅
- MDL Number:MFCD00036189
- Molecular Weight:647.89 g/mol
- UN:1170
- ADR:3,III
1 Options Available Below
Product Details & Documents
Proprietary, alcoholic and acidified formulation.
- Produces a brilliant cytoplasmic stain with three distinct color variations in a short amount of staining time
- Yields a very familiar cytoplasmic staining result
Part of the StatLab Select stain line.
Manufactured in USA.
Synonyms: Acid Red 87, Tetrabromofluorescein, 2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acid, 2,4,5,7-Tetrabromofluorescein, Eosin yellowish, Solvent red 43, Bromofluorescein, Bromo acid J. TS, XL, or XX, CI 45380, Bronze Bromo ES, Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-
- CAS Number:15086-94-9
- Formula:C₂₀H₈Br₄O₅
- MDL Number:MFCD00036189
- Molecular Weight:647.89 g/mol
- UN:1170
- ADR:3,III
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



