Thin-Walled PCR Tubes, 0.2 ml
470346-260
- Capacity:0.2 ml
- Description:PCR tube, thin-walled
- Cat. No.:470346-260
A thin-walled tube for efficient thermal transfer in PCR or other micro-reactions.
- PCR tube made of prime virgin polypropylene
- Transparent, soft and robust
- High light transmittance for optimal performance