United Scientific™ Ammeter, DC
Supplier: United Scientific Supplies
Certificates
About this item
Measure the current in DC circuits.
- DC ammeter mounted in black plastic case
- Scale length of 75 mm, with ±2.5% accuracy
- Features 5-way binding post and an external zero-setting screw
- Not protected against prolonged or significant overload
- Stackable for easy storage
DC ammeter mounted in a black plastic case. Meter scaled from 0 – 1A. Features 5-way binding post and an external zero-setting screw. Not protected against prolonged or significant overload. Stackable for convenient storage.
Frequently Bought Together