ContactUsLinkComponent
Innovating Science® Exothermic Reactions
Innovating Science® Exothermic Reactions
Catalog # 470343-428
Supplier:  Ward's Science
undefined
Innovating Science® Exothermic Reactions
Catalog # 470343-428
Supplier:  Ward's Science
Supplier Number:  IS2501
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Exothermic reactions
  • Cat. No.:
    470343-428
  • No. of uses:
    15
Specifications
About this item
Study the process of exothermic reactions.
  • Students will pass a bag around with an exothermic reaction occurring and feel the result of the reaction.
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2, HS-PS1-4, HS-PS3-2
  • Disciplinary core ideas: PS1.A, PS1.B, PS3.A
  • Cross cutting concepts include energy and matter. Structure and function, patterns
  • Engineering practices include developing and using models, analyzing and interpreting data, obtaining evaluating and communicating information
Allow students to see how chemistry can be used in a beneficial manner. Most students are probably familiar with heat packs, or hand warmers. Utilizing the release of chemical energy, these self-contained exothermic reactions are quite useful in cold environments.
In this activity, students will first investigate the temperature change of an exothermic reaction and then examine how the oxidation of iron, in a process similar to rusting, can be used to create the little bags of heat used to keep hands and feet warm. The kit contains enough materials for 15 groups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery: Kit includes instructional manual and SDS booklet.