- Anti-Static:+
- Closure type:With Zip Closure
- Garment application:Food,Paint and lacquer,Chemical protection,Services,Laboratory,Pharmaceutical
- Color:White
- Cat. No.:76538-710
Economical, lightweight protection from dirt, grease and light chemical splash in a global pattern.
- Global offering – great for multinational companies
- Global standardized testing and certification
- 4 sizing options simplify ordering and reduces inventory requirements (SM/MD, LG/XL, 2X/3X, 4X/5X)
- Exceeds ANSI 101-2014 size and label requirements for limited use and disposable coveralls
The MNSG 55gsm coverall offers protection against biological hazards, chemical splash, and dry particulates. Coveralls are suitable for applications such as: automotive, aerospace, paint booth operations, abatement/remediation, general maintenance, and infectious agents/biological hazards. These global pattern coveralls provide protection and comfort with a high Moisture Vapor Transmission Rate (MVTR) breathable fabric with excellent particle filtration efficiency. The chest label with testing and certification details is easily visible on each garment.
These 55gsm microporous film coveralls feature respirator fit hood, boots, protective storm flap over zipper, elastic wrists and ankles, serged seams, and elastic back.
Available in three configurations (MNSG428, MNSG417, and MNSG414) with four sizing options to simplify ordering and reduce inventory requirements.