Orders LinkContactUsLinkComponent
Vinnic Silver Oxide Button-Cell
Vinnic Silver Oxide Button-Cell
Silver oxide button cell battery
Catalog # 470346-472
Supplier #:  357F-C10VINNIC
Vinnic Silver Oxide Button-Cell
Catalog # 470346-472
Vinnic Silver Oxide Button-Cell
Vinnic Silver Oxide Button-Cell
Silver oxide button cell battery
Price
$7.76
Log in for availability
Pack of 10 Case of 100
Product Details & Documents
Button cell silver oxide coin battery in a convenient card of 10.
  • 0% mercury primary button cell
  • Silver oxide
  • Low internal resistance
  • Long service life
  • Top performance
Specifications
  • Description:
    Silver oxide button cell battery