Biox Microcentrifuge Tubes
Supplier: TWD Scientific
Certificates
About this item
These high quality boil proof microcentrifuge tubes are optically clear.
- Optically clear PP
- RNase, DNase, DNA, and PCR inhibitor free Boil proof cap
- Positive cap closure
The crystal clear microcentrigue tubes facilitate flawless visualization of samples and are ideal for high-temp applications.
Frequently Bought Together