ContactUsLinkComponent
MyBrix Handheld Refractometer, Mettler-Toledo
MyBrix Handheld Refractometer, Mettler-Toledo
Supplier:  Mettler Toledo
Certificates
undefined
MyBrix Handheld Refractometer, Mettler-Toledo
Supplier:  Mettler Toledo
Specifications
  • Catalog No:
  • 76595-302
  • 76547-950
  • Sample Volume:
  • 0.3 ml (min.)
  • 0.3 ml (min.)
  • Measurement Time:
  • 2 s
  • 2 s
  • Temperature Compensation Range:
  • 5...60 °C
  • 5...60 °C