Potassium sulfate 99-101%, meets analytical specification of Ph.Eur., Fluka™
BJ12658-1KG
:
- Pack type:Plastic bottle
- Size:6 x 1 kg
- Cat. No.:BJ12658-1KG
- Supplier no.:12658-1KG
- Assay (calculated to the dried substance):99.0 - 101.0 %
- Loss on drying (130°C):Max. 0.5 %
- Acidity or alkalinity:complying
- Calcium (Ca):Max. 50 ppm
- Iron (Fe):Max. 10 ppm
- Magnesium (Mg):Max. 20 ppm
- Sodium (Na):Max. 100 ppm
- Heavy metals (as Pb):Max. 0.001 %
- Chloride (Cl):Max. 20 ppm
- Appearance of the solution:complying
- Residual solvents:complying
:
:174.26 g/mol
:13,07759
:1.698 °C (1.013 hPa)
:1.067 °C
:2.66 g/cm³ (20 °C)
:Ambient
:231-915-5
:7778-80-5
:K₂SO₄
:MFCD00011388