ContactUsLinkComponent
Well Plate Tubes for Bond Elut SCX Plates, Agilent Technologies
Well Plate Tubes for Bond Elut SCX Plates, Agilent Technologies
Catalog # AG75507050
Well Plate Tubes for Bond Elut SCX Plates, Agilent Technologies
Catalog # AG75507050
Supplier Number:  75507050
Specifications
  • Ø:
    VersaPlate well plate tubes, SCX, 50 mg, 40 µm, PE frit (20 µm)
  • Volume:
    1.8 ml
  • Cat. No.:
    AG75507050
Specifications
About this item
Bond Elut SCX is a strong cation exchanger with a very low pKa and aromatic functionality.
  • Useful for compounds with cationic and nonpolar characteristics
  • Superior cleanup from a single sorbent
  • Very low pKa creates strong analyte interaction
Bond Elut SCX is a strong cation exchanger with a very low pKa. Although the pKa is similar to Bond Elut PRS, the presence of the benzene ring in the functional group increases the potential for nonpolar interactions. This nonpolar characteristic becomes important in ion exchange from aqueous systems.