ContactUsLinkComponent
Well Plate Tubes for Bond Elut C18 OH Plates, Agilent Technologies
Well Plate Tubes for Bond Elut C18 OH Plates, Agilent Technologies
Catalog # AG7551601C
Well Plate Tubes for Bond Elut C18 OH Plates, Agilent Technologies
Catalog # AG7551601C
Supplier Number:  7551601C
Specifications
  • Volume:
    1.8 ml
  • Description:
    VersaPlate well plate tubes, C18 OH, 100 mg, PE frit (20 µm)
  • Cat. No.:
    AG7551601C
Specifications
About this item
Bond Elut C18 OH enhances retention of basic compounds and promotes metabolite fractionation.
  • Silanol activity permits metabolite fractionation
  • Tight QC tolerances deliver batch-to-batch reproducibility
  • 50 Å pore size expands usage for higher molecular weight compounds
Bond Elut C18 OH is a nonendcapped version of the octadecyl bonded phases that enables the silanols on the silica surface to be more active. This low-load C18 phase has well-controlled silanol activity that permits the fractionation of metabolites and enhances retention of basic compounds compared to endcapped C18. The 150 Å pore size makes it a good choice for peptide extraction and higher molecular weight compounds.