Well Plate Tubes for Bond Elut PSA Plates, Agilent Technologies
Catalog # AG7552101C
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Specifications
- Volume:1.8 ml
- Description:VersaPlate well plate tubes, PSA, 100 mg, PE frit (20 µm)
- Cat. No.:AG7552101C