ContactUsLinkComponent
In-Line Filters for Thermo/Dionex LC Systems, Agilent Technologies
In-Line Filters for Thermo/Dionex LC Systems, Agilent Technologies
Catalog # AG8002-0806
undefined
In-Line Filters for Thermo/Dionex LC Systems, Agilent Technologies
Catalog # AG8002-0806
Supplier Number:  8002-0806
Specifications
  • Description:
    Agilent filter frit, titanium, 2 um, for inline filter in Thermo LPG-3400BM and DGP-3600BM pumps, similar to Dionex 6268.0036
  • Cat. No.:
    AG8002-0806
Specifications
About this item
High-quality in-line filters for selected Thermo Fisher Scientific HPLC systems.
  • In-line solvent filters, installed between the LC pump and injector, prevent blockages by removing particles from solvents before they reach the injector
  • Column inlet filters, positioned immediately before an LC column, remove particles from both the injection system and sample
Agilent offers in-line filters for Thermo Fisher Scientific HPLC systems, made from clean, high-quality stainless steel parts. These filters are designed to reduce column inlet frit contamination and blockage, which can increase backpressure and reduce efficiency, especially in microbore columns.