Specifications
- Description:Comparison LED microscope with tablet
- Cat. No.:470346-262
Specifications
About this item
A truly unique concept in microscopy!
- 8" LCD Android tablet with 1280×800 screen resolution
- Connection: WiFi; micro SD card
- WiFi (2.4 or 5 GHz) and Ethernet (RJ45)
- MotiConnect App
- Connect five WiFi devices at one time
The M3-F forensic microscope offers professional features and detachable 8" tablet at a highly competitive price point. Use the M3-F's revolutionary technology to compare images in both micro and macro environments. The M3's dedicated macro lens, with a large working distance, allows you to view not only bullets but also larger more bulky, 'evidence' items.
Swift's powerful optical system allows for images to be seen either 100% from the left microscope, 100% from the right, side-by-side, or overlapping. The M3-F is the first and only comparison microscope at this price level that can achieve overlapping of images.
Combining the features of Swift's popular M3-F's advanced comparison microscope with an Adroid based tablet system offers unlimited possibilities.