Order Entry
United States
ContactUsLinkComponent
Motic Swift Line M30TZ Zoom Stereo Microscope
Motic Swift Line M30TZ Zoom Stereo Microscope
Catalog # 470346-270
Supplier:  Swift Instruments
undefined
Motic Swift Line M30TZ Zoom Stereo Microscope
Catalog # 470346-270
Supplier:  Swift Instruments
Supplier Number:  M30TZ-SM99CL

Specifications

  • Description:
    Zoom stereo microscope
  • Cat. No.:
    470346-270

Specifications

About this item

Versatile and durable, ideal for high school classrooms and beyond.

  • Paired widefield eyepieces are 45° inclined, with diopter adjustment
  • Interpupillary adjustment from 55 to 75 mm
  • Interlocked eyepiece tubes with diopter for increased focus acuity
  • Viewing head rotates 360°, locks in place by tightening thumb screw

The M30TZ is ideal for laboratory, industrial and educational use. This stereo microscope features zoom objectives with a continuous magnification range of 7.5× to 40×, offering added versatility that is not an option with fixed power stereo microscopes. Trinocular Port accepts video adapter.